Human Recombinant 14-3-3 epsilon (His tag) (from Baculovirus-Insect Cells)
Supplier: Sino Biological
Y75-30H-100 Y75-30H-1000 Y75-30H-20
77565-580EA 1971.84 CAD
77565-580 77565-582 77565-584
Human Recombinant 14-3-3 epsilon (His tag) (from Baculovirus-Insect Cells)
Proteins and Peptides
Recombinant full length human 14-3-3 epsilon was expressed by baculovirus in Sf9 insect cells using an N-terminal His tag.
  • Shipping temperature: Dry ice