Mouse Recombinant 14-3-3 epsilon (His tag) (from Baculovirus-Insect Cells)
Supplier: Sino Biological
Y75-30BH-100 Y75-30BH-1000 Y75-30BH-20
77565-568EA 1971.84 CAD
77565-568 77565-570 77565-572
Mouse Recombinant 14-3-3 epsilon (His tag) (from Baculovirus-Insect Cells)
Proteins and Peptides
Recombinant full length mouse 14-3-3 epsilon was expressed by baculovirus in Sf9 insect cells using an N-terminal His tag.
  • Shipping temperature: Dry ice