Human Recombinant 14-3-3 alpha/beta (GST tag) (from E. coli)

Supplier: Sino Biological

Y71-30G-100 Y71-30G-1000
77565-550EA 1102.84 CAD
77565-550 77565-552
Human Recombinant 14-3-3 alpha/beta (GST tag) (from E. coli)
Proteins and Peptides

Recombinant full length human 14-3-3 alpha/beta was expressed in E. coli cells using a N-terminal GST tag.


  • Shipping temperature: Dry ice

New Product

Order Now

SPECIFICATIONS

Protein/peptide name YWHAB
Protein synonyms HS1|GW128|KCIP-1|YWHAA|HEL-S-1
Environmentally Preferable
Protein/peptide type Recombinant
Species Human
Source E. coli
Conjugation Unconjugated
Formulation Supplied as sterile 50 mM Tris-HCl, pH 7.5, 50 - 300 mM NaCl, 10 mM glutathione, 0.1 mM EDTA, 0.25 - 1.0 mM DTT, 0 - 0.1 mM PMSF, 10 - 25% glycerol
Molecular weight ~54 kDa
Storage conditions Store product at –70 °C. For optimal storage, aliquot target into smaller quantities after centrifugation and store at recommended temperature. For most favorable performance, avoid repeated handling and multiple freeze/thaw cycles.

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR