Mouse Recombinant WEE2 (GST tag), Active (from Baculovirus-Insect Cells)
Supplier: Sino Biological
W06-10G-10 W06-10G-50 W06-10G-500
77565-454EA 549.84 CAD
77565-454 77565-456 77565-458
Mouse Recombinant WEE2 (GST tag), Active (from Baculovirus-Insect Cells)
Proteins and Peptides
Recombinant full length mouse WEE2 was expressed by baculovirus in Sf9 insect cells using an N-terminal GST tag.
  • Shipping temperature: Dry ice