Human Recombinant VRK2 (GST tag), Active (from Baculovirus-Insect Cells)
Supplier: Sino Biological
V02-11G-50 V02-11G-500
77565-374EA 1845.44 CAD
77565-374 77565-376
Human Recombinant VRK2 (GST tag), Active (from Baculovirus-Insect Cells)
Proteins and Peptides
Recombinant human VRK2 (1-375) was expressed by baculovirus in Sf9 insect cells using an N-terminal GST tag.
  • Shipping temperature: Dry ice