Human Recombinant USP20 (GST tag), Active (from Baculovirus-Insect Cells)
Supplier: Sino Biological
U520-381G-10
U520-381G-50
U520-381G-500
77565-302EA
518.24
CAD
77565-302
77565-304
77565-306
Human Recombinant USP20 (GST tag), Active (from Baculovirus-Insect Cells)
Proteins and Peptides
Recombinant human USP20 (120-end) was expressed by baculovirus in Sf9 insect cells using an N-terminal GST tag. The USP20 gene accession number is NM_006676.
- Shipping temperature: Dry ice