Human Recombinant Tau-441, BRSK1-phosphorylated (from E. coli)
Supplier: Sino Biological
T08-50N-100 T08-50N-1000 T08-50N-20
77564-456EA 1608.44 CAD
77564-456 77564-458 77564-460
Human Recombinant Tau-441, BRSK1-phosphorylated (from E. coli)
Proteins and Peptides
Recombinant human tag-free Tau-441 was co-expressed with BRSK1 in E. coli cells. The Tau-441 was phosphorylated by BRSK1 in vivo and in vitro prior to the final chromatography purification.
  • Shipping temperature: Dry ice