Human Recombinant Tau-441, PKA-phosphorylated (from E. coli)
Supplier: Sino Biological
T08-50LN-100 T08-50LN-1000 T08-50LN-20
77564-450EA 1608.44 CAD
77564-450 77564-452 77564-454
Human Recombinant Tau-441, PKA-phosphorylated (from E. coli)
Proteins and Peptides
Recombinant human tag-free Tau-441 was co-expressed with PKAc alpha in E. coli cells. The Tau-441 protein was phosphorylated by PKAc alpha in vivo and in vitro prior to the final chromatography purification.
  • Shipping temperature: Dry ice