Mouse Recombinant TGFBR2 (GST tag), Active (from Baculovirus-Insect Cells)
Supplier: Sino Biological
T08-11BG-10 T08-11BG-50 T08-11BG-500
77564-416EA 549.84 CAD
77564-416 77564-418 77564-420
Mouse Recombinant TGFBR2 (GST tag), Active (from Baculovirus-Insect Cells)
Proteins and Peptides
Recombinant mouse TGFBR2 (215-end) was expressed by baculovirus in Sf9 insect cells using an N-terminal GST tag.
  • Shipping temperature: Dry ice