Mouse Recombinant TGFBR1 (ALK5, GST tag), Active (from Baculovirus-Insect Cells)
Supplier: Sino Biological
T07-11BG-50 T07-11BG-500
77564-374EA 1845.44 CAD
77564-374 77564-376
Mouse Recombinant TGFBR1 (ALK5, GST tag), Active (from Baculovirus-Insect Cells)
Proteins and Peptides
Recombinant mouse TGFBR1 (ALK5) (148-end) was expressed by baculovirus in Sf9 insect cells using an N-terminal GST tag.
  • Shipping temperature: Dry ice