Human Recombinant TIE2 (GST tag), Active (from Baculovirus-Insect Cells)
Supplier: Sino Biological
T04-11G-50 T04-11G-500
77564-268EA 1845.44 CAD
77564-268 77564-270
Human Recombinant TIE2 (GST tag), Active (from Baculovirus-Insect Cells)
Proteins and Peptides
Recombinant human TIE 2 (771-end) was expressed by baculovirus in Sf9 insect cells using an N-terminal GST tag.
  • Shipping temperature: Dry ice