Human Recombinant STAT5B (GST tag) (from Baculovirus-Insect Cells)
Supplier: Sino Biological
S56-54BG-100 S56-54BG-1000 S56-54BG-20
77564-194EA 1624.24 CAD
77564-194 77564-196 77564-198
Human Recombinant STAT5B (GST tag) (from Baculovirus-Insect Cells)
Proteins and Peptides
Recombinant full length human STAT5B was expressed by baculovirus in Sf9 insect cells using an N-terminal GST tag.
  • Shipping temperature: Dry ice