Human Recombinant NDR (GST tag), Unactive (from Baculovirus-Insect Cells)
Supplier: Sino Biological
S49-14G-100 S49-14G-1000
77564-086EA 1292.44 CAD
77564-086 77564-088
Human Recombinant NDR (GST tag), Unactive (from Baculovirus-Insect Cells)
Proteins and Peptides
Recombinant full length human NDR was expressed by baculovirus in Sf9 insect cells using an N-terminal GST tag.
  • Shipping temperature: Dry ice