Human Recombinant SIRT1 (His tag), Active (from Baculovirus-Insect Cells)
Supplier: Sino Biological
S35-31H-50 S35-31H-500
77563-910EA 2050.84 CAD
77563-910 77563-912
Human Recombinant SIRT1 (His tag), Active (from Baculovirus-Insect Cells)
Proteins and Peptides
Recombinant human SIRT1 (193-end) was expressed by baculovirus in Sf9 insect cells using an N-terminal His tag.
  • Shipping temperature: Dry ice