Human Recombinant SIRT1 (GST tag), Active (from Baculovirus-Insect Cells)
Supplier: Sino Biological
S35-31G-10 S35-31G-50 S35-31G-500
77563-902EA 660.44 CAD
77563-902 77563-904 77563-906
Human Recombinant SIRT1 (GST tag), Active (from Baculovirus-Insect Cells)
Proteins and Peptides
Recombinant human SIRT1 (193-end) was expressed by baculovirus in Sf9 insect cells using an N-terminal GST tag.
  • Shipping temperature: Dry ice