Human Recombinant SOD3 (GST tag) (from Baculovirus-Insect Cells)
Supplier: Sino Biological
S28-31G-100 S28-31G-1000 S28-31G-20
77563-762EA 1087.04 CAD
77563-762 77563-764 77563-766
Human Recombinant SOD3 (GST tag) (from Baculovirus-Insect Cells)
Proteins and Peptides
Recombinant full length human SOD3 was expressed by baculovirus in Sf9 insect cells using an N-terminal GST tag.
  • Shipping temperature: Dry ice