Human Recombinant Alpha Synuclein Monomer (Type 1) (from E. coli)

Supplier: Sino Biological

S22-371N-100 S22-371N-200 S22-371N-500
77563-690EA 718.9 CAD
77563-690 77563-692 77563-694
Human Recombinant Alpha Synuclein Monomer (Type 1) (from E. coli)
Proteins and Peptides

Recombinant full length tag-free human Alpha-synuclein Protein Monomer (Type 1) was expressed in E. coli cells.


  • Shipping temperature: Dry ice

Order Now

SPECIFICATIONS

Protein/peptide name SNCA
Protein synonyms PARK1|PARK4|NACP|PD1
Environmentally Preferable
Protein/peptide type Recombinant
Species Human
Source E. coli
Conjugation Unconjugated
Formulation Supplied as sterile PBS, pH 7.4.
Molecular weight 14.5 kDa
Storage conditions Store product at –70 °C. For optimal storage, aliquot target into smaller quantities after centrifugation and store at recommended temperature. For most favorable performance, avoid repeated handling and multiple freeze/thaw cycles.

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR