Human Recombinant SGK2 (GST tag), Active (from Baculovirus-Insect Cells)
Supplier: Sino Biological
S07-10G-500 S07-10G-50
77563-468EA 4168.04 CAD
77563-468 77563-466
Human Recombinant SGK2 (GST tag), Active (from Baculovirus-Insect Cells)
Recombinant full length human SGK2 was expressed by baculovirus in Sf9 insect cells using an N-terminal GST tag.
  • Shipping temperature: Dry ice