Human Recombinant SGK1 (GST tag), Unactive (from Baculovirus-Insect Cells)
Supplier: Sino Biological
S06-14G-100 S06-14G-1000 S06-14G-20
77563-450EA 1292.44 CAD
77563-450 77563-452 77563-454
Human Recombinant SGK1 (GST tag), Unactive (from Baculovirus-Insect Cells)
Proteins and Peptides
Recombinant full length human SGK1 was expressed by baculovirus in Sf9 insect cells using an N-terminal GST tag.
  • Shipping temperature: Dry ice