Human Recombinant DGKE (GST tag), Active (from Baculovirus-Insect Cells)
Supplier: Sino Biological
D24-10G-50 D24-10G-500 D24-10G-10
77556-108EA 2208.84 CAD
77556-108 77556-110 77556-106
Human Recombinant DGKE (GST tag), Active (from Baculovirus-Insect Cells)
Proteins and Peptides
Recombinant full length human DGKE was expressed by baculovirus in Sf9 insect cells using an N-terminal GST tag.
  • Shipping temperature: Dry ice