Dog Recombinant DGKA (His tag), Active (from Baculovirus-Insect Cells)
Supplier: Sino Biological
D21-10DH-10 D21-10DH-50 D21-10DH-500
77556-048EA 628.84 CAD
77556-048 77556-050 77556-052
Dog Recombinant DGKA (His tag), Active (from Baculovirus-Insect Cells)
Proteins and Peptides
Recombinant full length dog DGKA was expressed by baculovirus in Sf9 insect cells using an N-terminal His tag.
  • Shipping temperature: Dry ice