Mouse Recombinant DGKA (GST tag), Active (from Baculovirus-Insect Cells)
Supplier: Sino Biological
D21-10CG-10 D21-10CG-50 D21-10CG-500
77556-036EA 628.84 CAD
77556-036 77556-038 77556-040
Mouse Recombinant DGKA (GST tag), Active (from Baculovirus-Insect Cells)
Proteins and Peptides
Recombinant full length mouse DGKA was expressed by baculovirus in Sf9 insect cells using an N-terminal GST tag.
  • Shipping temperature: Dry ice