Human Recombinant DAPK1 (GST tag), Active (from Baculovirus-Insect Cells)
Supplier: Sino Biological
D01-11G-50 D01-11G-500
77555-910EA 1845.44 CAD
77555-910 77555-912
Human Recombinant DAPK1 (GST tag), Active (from Baculovirus-Insect Cells)
Proteins and Peptides
Recombinant human DAPK1 (1-363) was expressed by baculovirus in Sf9 insect cells using an N-terminal GST tag.
  • Shipping temperature: Dry ice