Human Recombinant CyclinO (GST tag) (from Baculovirus-Insect Cells)
Supplier: Sino Biological
C85-30G-100 C85-30G-1000 C85-30G-20
77555-878EA 1292.44 CAD
77555-878 77555-880 77555-882
Human Recombinant CyclinO (GST tag) (from Baculovirus-Insect Cells)
Proteins and Peptides
Recombinant full length human Cyclin O was expressed by baculovirus in Sf9 insect cells using an N-terminal GST tag.
  • Shipping temperature: Dry ice