Human Recombinant CyclinD2 (GST tag) (from Baculovirus-Insect Cells)
Supplier: Sino Biological
C81-30B2G-100 C81-30B2G-1000 C81-30B2G-20
77555-832EA 1292.44 CAD
77555-832 77555-834 77555-836
Human Recombinant CyclinD2 (GST tag) (from Baculovirus-Insect Cells)
Proteins and Peptides
Recombinant full length human CyclinD2 was expressed by baculovirus in Sf9 insect cells using an N-terminal GST tag.
  • Shipping temperature: Dry ice