Human Recombinant FMS (GST tag), Active (from Baculovirus-Insect Cells)
Supplier: Sino Biological
C74-11G-50 C74-11G-500
77555-800EA 1845.44 CAD
77555-800 77555-802
Human Recombinant FMS (GST tag), Active (from Baculovirus-Insect Cells)
Proteins and Peptides
Recombinant human FMS (539-end) was expressed by baculovirus in Sf9 insect cells using a N-terminal GST tag.
  • Shipping temperature: Dry ice