Human Recombinant CDKL4 (GST tag) (from Baculovirus-Insect Cells)
Supplier: Sino Biological
C69-30G-100 C69-30G-1000 C69-30G-20
77555-782EA 1292.44 CAD
77555-782 77555-784 77555-786
Human Recombinant CDKL4 (GST tag) (from Baculovirus-Insect Cells)
Proteins and Peptides
Recombinant full length human CDKL4 was expressed by baculovirus in Sf9 insect cells using an N-terminal GST tag.
  • Shipping temperature: Dry ice