Human Recombinant CK1A1 (E42C, GST tag), Active (from Baculovirus-Insect Cells)
Supplier: Sino Biological
C64-12CG-10 C64-12CG-50 C64-12CG-500
77555-736EA 534.04 CAD
77555-736 77555-738 77555-740
Human Recombinant CK1A1 (E42C, GST tag), Active (from Baculovirus-Insect Cells)
Proteins and Peptides
Recombinant full length human CK1A1 (E42C) was expressed by baculovirus in Sf9 insect cells using an N-terminal GST tag.
  • Shipping temperature: Dry ice