Rat Recombinant CAMKK1 (GST tag), Active (from Baculovirus-Insect Cells)
Supplier: Sino Biological
C17-18G-500 C17-18G-10 C17-18G-50
77555-168EA 4168.04 CAD
77555-168 77555-164 77555-166
Rat Recombinant CAMKK1 (GST tag), Active (from Baculovirus-Insect Cells)
Proteins and Peptides
Recombinant full length rat CAMKK1 was expressed by baculovirus in Sf9 insect cells using an N-terminal GST tag.
  • Shipping temperature: Dry ice