Human Recombinant BTK (C481R, His tag), Active (from Baculovirus-Insect Cells)
Supplier: Sino Biological
B10-12GH-10 B10-12GH-50 B10-12GH-500
77554-796EA 534.04 CAD
77554-796 77554-798 77554-800
Human Recombinant BTK (C481R, His tag), Active (from Baculovirus-Insect Cells)
Proteins and Peptides
Recombinant full length human BTK (C481R) was expressed by baculovirus in Sf9 insect cells using an N-terminal His tag.
  • Shipping temperature: Dry ice