Human Recombinant EML4-ALK (Eex20Aex20, GST tag), Active (from Baculovirus-Insect Cells)
Supplier: Sino Biological
A19-19HG-10 A19-19HG-50 A19-19HG-500
77554-300EA 613.04 CAD
77554-300 77554-302 77554-304
Human Recombinant EML4-ALK (Eex20Aex20, GST tag), Active (from Baculovirus-Insect Cells)
Proteins and Peptides
Recombinant human fusion protein EML4 (1-580) - ALK (1058-end) was expressed by baculovirus in Sf9 insect cells using an N-terminal GST tag.
  • Shipping temperature: Dry ice