Human Recombinant ALK (GST tag), Active (from Baculovirus-Insect Cells)
Supplier: Sino Biological
A19-11G-10 A19-11G-50 A19-11G-500
77554-168EA 549.84 CAD
77554-168 77554-170 77554-172
Human Recombinant ALK (GST tag), Active (from Baculovirus-Insect Cells)
Proteins and Peptides
Recombinant human ALK (1060-end) was expressed by baculovirus in Sf9 insect cells using an N-terminal GST tag.
  • Shipping temperature: Dry ice