Human Recombinant ABL1 (His tag), Active (from Baculovirus-Insect Cells)
Supplier: Sino Biological
A03-18H-50 A03-18H-500
77553-842EA 1845.44 CAD
77553-842 77553-844
Human Recombinant ABL1 (His tag), Active (from Baculovirus-Insect Cells)
Proteins and Peptides
Recombinant human ABL1 (27-end) was expressed by baculovirus in Sf9 insect cells using an N-terminal His tag.
  • Shipping temperature: Dry ice