Human Recombinant ABL1 (K290R, His tag) (from Baculovirus-Insect Cells)
Supplier: Sino Biological
A03-16NG-100 A03-16NG-1000 A03-16NG-20
77553-836EA 1292.44 CAD
77553-836 77553-838 77553-840
Human Recombinant ABL1 (K290R, His tag) (from Baculovirus-Insect Cells)
Proteins and Peptides
Recombinant human ABL1 (K290R) (27-end) was expressed by baculovirus in Sf9 insect cells using an N-terminal His tag. ABL1 (K290R) is a kinase dead mutant.
  • Shipping temperature: Dry ice