Human Recombinant ABL1 (Y245F Y412F, His tag), Active (from Baculovirus-Insect Cells)
Supplier: Sino Biological
A03-12QH-10 A03-12QH-50 A03-12QH-500
77553-830EA 534.04 CAD
77553-830 77553-832 77553-834
Human Recombinant ABL1 (Y245F Y412F, His tag), Active (from Baculovirus-Insect Cells)
Proteins and Peptides
Recombinant human ABL1 (Y245F Y412F) (27-end) was expressed by baculovirus in Sf9 insect cells using an N-terminal His tag.
  • Shipping temperature: Dry ice