Human Recombinant ABL1 (F317I, GST tag), Active (from Baculovirus-Insect Cells)

Supplier: Sino Biological

A03-12JG-10 A03-12JG-50 A03-12JG-500
77553-800EA 534.04 CAD
77553-800 77553-802 77553-804
Human Recombinant ABL1 (F317I, GST tag), Active (from Baculovirus-Insect Cells)
Proteins and Peptides

Recombinant human ABL1 (F317I) (27-end) was expressed by baculovirus in Sf9 insect cells using an N-terminal GST tag.


  • Shipping temperature: Dry ice

Order Now

SPECIFICATIONS

Protein/peptide name ABL1
Protein synonyms c-ABL|ABL|JTK7|bcr/abl|p150|v-abl|c-ABL1
Environmentally Preferable
Protein/peptide type Recombinant
Species Human
Source Baculovirus (Insect cells)
Conjugation Unconjugated
Formulation Supplied as sterile 50 mM Tris-HCl, pH 7.5, 50 - 300 mM NaCl, 10 mM glutathione, 0.1 mM EDTA, 0.25 - 1.0 mM DTT, 0 - 0.1 mM PMSF, 10 - 25% glycerol
Molecular weight ~160 kDa
Storage conditions Store product at –70 °C. For optimal storage, aliquot target into smaller quantities after centrifugation and store at recommended temperature. For most favorable performance, avoid repeated handling and multiple freeze/thaw cycles.
Tested applications Kinase Assay

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR