Human Recombinant ABL1 (Y253H, His tag), Active (from Baculovirus-Insect Cells)
Supplier: Sino Biological
A03-12GH-10 A03-12GH-50 A03-12GH-500
77553-782EA 534.04 CAD
77553-782 77553-784 77553-786
Human Recombinant ABL1 (Y253H, His tag), Active (from Baculovirus-Insect Cells)
Proteins and Peptides
Recombinant human ABL1 (Y253H) (27-end) was expressed by baculovirus in Sf9 insect cells using an N-terminal His tag.
  • Shipping temperature: Dry ice