Human Recombinant ABL1 (Q252H, GST tag), Active (from Baculovirus-Insect Cells)
Supplier: Sino Biological
A03-12FG-10 A03-12FG-50 A03-12FG-500
77553-776EA 534.04 CAD
77553-776 77553-778 77553-780
Human Recombinant ABL1 (Q252H, GST tag), Active (from Baculovirus-Insect Cells)
Proteins and Peptides
Recombinant human ABL1 (Q252H) (27-end) was expressed by baculovirus in Sf9 insect cells using an N-terminal GST tag.
  • Shipping temperature: Dry ice