ProKlenz® Foam High Performance Alkaline Cleaner

Supplier: Steris
143102WR 143105WR
77531-588EA 2992.68 CAD
77531-588 77531-594
ProKlenz® Foam High Performance Alkaline Cleaner
Cleaning Agents Detergents and Surface Cleansers
All-purpose foam cleaner.

  • Standardization - Globally available product enables harmonized cleaning protocols across sites
  • Sustainability - Free rinsing formula minimizes utility consumption and detergent residue
  • Effectiveness - Alkaline compatible surfactant system cleans efficiently with one high foaming product
  • Efficiency - A high foaming system and low use-concentration creates a robust foam profile that promotes cost-effective cleaning and minimizes cleaning time
  • Quality - Manufactured within an FDA registered plant, ISO 9001:2015 and ISO 13485:2016 compliant, to meet the highest standard in manufacturing and quality assurance

When used as directed with suitable foaming spray equipment, ProKlenz® FOAM Cleaner can be applied on internal and external processing equipment constructed from common types of metals and plastics. Emulsification and dispersion properties create an effective foam profile for cleaning fats, oils, and organic soils.

ProKlenz® Foam High Performance Alkaline Cleaner is an engineered all-purpose cleaner to accommodate the pharmaceutical, biotechnology, cosmetic, and nutraceutical industries worldwide. ProKlenz FOAM Cleaner has a high foam profile, even at a low concentration, for improved cleaning over existing foam products. The free rinsing formula and surfactant system minimize utility consumption and promote cost-effective cleaning processes.

CAS: 1310-58-3; 68855-69-6; 1312-76-1; 68479-09-4; 34590-94-8; 68439-57-6; 68439-46-3
UN 1814
Hazard class: 8
Packing group: II

Caution: Danger, GHS05; GHS07.
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR