Vesphene® III se Phenolic Disinfectants
Supplier: Steris
647505 647501 647508
76303-114EA 789.87 CAD
76303-114 76303-116 76303-152
Vesphene® III se Phenolic Disinfectants
Cleaning Agents Disinfectants
STERIS' Vesphene® III se Phenolic disinfectant is a liquid concentrate specifically formulated for the disinfection of hard surfaces. The use-dilution can be applied manually or sprayed.
  • Efficacy - Phenolic active ingredients provide broad spectrum antimicrobial efficacy
  • Productivity - Formulation includes surfactants for surface cleaning while disinfecting
  • Flexibility - Ideal for use in a contamination control rotation
  • Reliability - Complete lot traceability
  • Assurance - Each lot documented by Certificate of Analysis
STERIS' Vesphene® III se disinfectant is specifically formulated for the routine disinfection of hard, non-porous surfaces. At a use dilution of rate of 0.8% v/v (1:128), the Vesphene® III disinfectants kill a broad spectrum of common bacteria and fungi. The alkaline Vesphene® III disinfectant formulation can be used alone or in rotation with the LpH®® III disinfectants. These products have an extensive documentation package to meet your validation objectives and are supported by the STERIS Technical Services team for individualized application and validation assistance.