StarFish™ Glassware Condensers
Supplier: Heidolph NA, LLC
015890760 015890770 015890780 015890790 015890800 015890810 015890760CA 015890770CA 015890780CA 015890790CA 015890800CA 015890810CA
CA11007-832EA 176.26 CAD
CA11007-832 CA11007-834 CA11007-836 CA11007-838 CA11007-840 CA11007-842 CA11009-146 CA11009-148 CA11009-150 CA11009-152 CA11009-154 CA11009-156
StarFish™ Glassware Condensers
Condensers
Condensers can be used for all standard distillations.
  • Different types of connections available
  • Available in various lengths/cone sizes