Erlenmeyer Flasks, Polycarbonate, Flat Bottom
Supplier: NEST SCIENTIFIC
786001 786011 785001
77508-880CS 424.93 CAD
77508-880 77508-882 77508-864
Erlenmeyer Flasks, Polycarbonate, Flat Bottom
Flasks Erlenmeyer Flasks
PC Erlenmeyer flasks are manufactured in 100000 grade cleaning room with USP 6 grade raw materials.
  • PC Bottle and HDPE cap
  • Vent caps with 0.2 μm hydrophobic filters for gas exchange without contamination
  • Individually packaged in sterile bags
  • Sterilized by E-beam, SAL= 10⁻⁶
  • Non-pyrogenic, DNase/Rnase free
PC Bottle and HDPE cap, vent caps with 0.2 µm hydrophobic filters for gas exchange without contamination. Ideal for shaker culture applications.
Certifications: ISO 9001 and 13485 certified.