PYREX® Flat Bottom Culture Tubes, Rimless, Corning
Supplier: Corning
9850-25 9850-25XX
60856-130CS 470.4 CAD
60856-130 60856-132 60856-134
PYREX® Flat Bottom Culture Tubes, Rimless, Corning
Tubes Culture Tubes
Flat-bottom culture tubes are ideal for plant tissue and general culture work.
Two-position polypropylene closures are sold separately. Position closure open for gas exchange or closed for humidified environment. Internal drip ring minimizes contamination.
The flat bottom facilitates use with agar slants. Walls and bottoms are of uniform thickness. The tubes are rimless for greater convenience in plugging and rack storage.
Ordering information: Tubes and closures sold separately.