KIMAX® Culture Tubes, Reusable, with Screw Cap, Kimble Chase
Supplier: DWK Life Sciences (KIMBLE)
45066-38200 45066-16150 45066A-25200 45066-13100 45066-16100 45066-16125 45066-20125 45066-20150 45066-25150 45066-25200 45066A-13100 45066A-16100 45066A-16125 45066A-16150 45066A-20125 45066A-20150 45066A-25150
89001-476CS 968.95 CAD
89001-476 89000-860 89000-862 89001-462 89001-464 89001-466 89001-468 89001-470 89001-472 89001-474 89001-478 89001-480 89001-482 89001-484 89001-486 89001-488 89001-490
KIMAX® Culture Tubes, Reusable, with Screw Cap, Kimble Chase
Tubes Culture Tubes
Tubes with uniform wall thickness for maximum heat transfer and chemical resistance.
Tubes feature a marking spot and a threaded phenolic screw cap that can be loosened for autoclaving. Tubes are available with rubber-lined caps or PTFE-faced rubber-lined caps.
Certifications: Tubes with rubber-lined caps are designed from ASTM Specification E982, Type VI, Class A requirements. Tubes with PTFE-faced rubber-lined caps are designed from ASTM Specification E982, Type VI, Class B requirements.
Ordering information: Cap is supplied unattached.