PYREX® Culture Tube, Screw Cap, Corning
Supplier: Corning
9825-25X 9825-22 9825-13 9825-16 9825-16X 9825-16XX 9825-20 9825-20X 9825-38 9825-25
60827-205CS 1346.31 CAD
60827-205 89091-462 60827-023 60827-045 60827-067 60827-089 60827-125 60827-147 60827-207 89090-930
PYREX® Culture Tube, Screw Cap, Corning
Tubes Culture Tubes
Furnished with deep-form phenolic caps to facilitate handling and sealing after autoclaving.
Caps are fabricated from a special formula resistant to the effects of temperature and steam in autoclaving. White enamel marking spot allows excellent surface for pencil notations. Pencil marks are easily erased.
For phenolic caps, see Cat. No. 9999.
Ordering information: Choose from tubes with rubber-lined screw caps or fluoropolymer resin-faced rubber-lined screw caps.
Order Now
SPECIFICATIONS
Environmentally Preferable Low Manufacturing Impact
Environmental certification I-RECs, ISO 14001 - Monterrey