PYREX® Test Tubes with Beaded Rim, Corning
Supplier: Corning
9800-10 9800-12 9800-13 9800-15 9800-16 9800-18 9800-20 9800-25 9800-25X
60815-024CS 973.19 CAD
60815-024 60815-046 60815-068 60815-104 60815-126 60815-148 60815-160 60815-228 60815-240
PYREX® Test Tubes with Beaded Rim, Corning
Tubes Test Tubes
These tubes are made from special tubing to give the optimum wall thickness.
They are well annealed, resistant to heat, and chemically stable. Rims are fire-polished.
Order Now
SPECIFICATIONS
Environmentally Preferable Low Manufacturing Impact
Environmental certification I-RECs, ISO 14001 - Monterrey