Prelabeled Culture Tubes, Disposable, Borosilicate Glass, Kimble Chase
Supplier: DWK Life Sciences (KIMBLE)
60B12BZW 60A10BZW 60C13BZW 60A10BZG 60A10BZL 60B12BZG
60826-110CS 360.94 CAD
60826-110 60600-316 60600-334 89236-684 89236-686 89236-710
Prelabeled Culture Tubes, Disposable, Borosilicate Glass, Kimble Chase
Tubes Culture Tubes
Writing placed on the labels of these tubes stays permanently, making them ideal for use in cell washers and water baths.
Fused-on labels provide ideal surface for marking. Eliminate many problems resulting from smeared or illegible identification of test samples. Size of label on 10 and 12 mm Ext.Ø tubes is 1.9×3.5 cm (³/₄×1³/₈"). Size of label on 13 and 16 mm Ext.Ø tubes is 2.5×5.1cm (1×2").
Ordering information: To order tubes with green, brown, or gray labels, or to order blue or yellow labels on 13×100 mm and 16×100 mm tubes, contact your Avantor sales representative.