Leucosep™ Centrifuge Tubes, Greiner Bio-One
Supplier: Greiner Bio-One
163290 163290P 227290 227290P 227288 163288
89048-932CS 2761.72 CAD
89048-932 89048-934 89048-936 89048-938 89136-192 89218-662
Leucosep™ Centrifuge Tubes, Greiner Bio-One
Clinical Diagnostic Supplies Clinical Sample Collection/Preparation Blood Sample Collection/Preparation
Leucosep™ tubes have been developed for the optimal separation of lymphocytes and peripheral mononuclear cells from human whole blood and bone marrow by means of density gradient centrifugation.
  • Transparent, porous barrier of high-grade polyethylene
  • Sterile tubes
  • Tubes conical bottom, graduated, made of polypropylene
  • Designed for single-use only
The key feature of these tubes is the transparent, porous, biologically inert barrier constructed of high-grade polyethylene
Sterile tubes offer improved yields and recoveries, higher purity enrichments,More Product Information