CELLSTAR® Light-Impermeable Centrifuge Tubes, Polypropylene, Sterile, Greiner Bio-One
Supplier: Greiner Bio-One
188280 188283
82051-624CS 580.1 CAD
82051-624 82051-626 82051-628 82051-630
CELLSTAR® Light-Impermeable Centrifuge Tubes, Polypropylene, Sterile, Greiner Bio-One
Tubes Centrifuge Tubes
Tubes offer a substantial reduction or elimination of light penetration to contents, enabling their use for storage of light-sensitive materials and applications for light-sensitive reactions.
  • Polypropylene construction
  • Amber tint protects light-sensitive materials
  • Molded graduations
  • Clear marking space for sample identification
Constructed of amber pigmented polypropylene, these conical-bottom tubes feature molded graduations, a clear marking space for sample identification, and blue screw caps. All tubes are supplied sterile.