CELLSTAR® Centrifuge Tubes, Polypropylene, Sterile, Greiner Bio-One
Supplier: Greiner Bio-One
188281 227270 227281 188261 188271 210261 210270 227261 227285 188271-TRI 227261-TRI
82050-280CS 815.54 CAD
82050-280 82050-282 82050-348 82050-350 82050-276 82050-278 82050-320 82050-322 82050-346 82050-352 75799-844 75799-846
CELLSTAR® Centrifuge Tubes, Polypropylene, Sterile, Greiner Bio-One
Tubes Centrifuge Tubes
These conical-bottom, sterile centrifuge tubes are made from ClearChoice™ polypropylene with excellent optical clarity and high RCF ratings.
  • Sterile
  • Free of RNase, DNase, and human DNA
  • Non-pyrogenic
  • Available with or without a skirt
  • Polypropylene construction
Tubes feature graduations and an ID field printed on the side.
Ordering information: Supplied with HDPE flat-top screw caps.